Learn More
MilliporeSigma™ Spill Kit, Mercury, SpillSolv
Complete kit includes container, reagents, gloves, collector pan and brush, disposal bags, spray bottle and treatment guide
Supplier: MilliporeSigma™ SX13501
Specifications
Mercury Spill Kit | |
Poly container, 750g shaker bottle SpillSolv mercury immobilizer, 180g SpillSolv mercury vapor suppressant, collector pan and brush, three 14 x 20 in. x 4 mil H.D. disposal bags, goggles, gloves, disposal bags, scoops, sponge, and guide |