Learn More
MilliporeSigma™ Novagen™ Veggie BL21 (DE3) Singles™ Competent Cells
Protease-deficient hosts (BL21) for protein expression; Manufactured free of animal-derived media and components
$302.00 - $537.00
Specifications
Content And Storage ≤-70°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
71-252-3
View Documents
MilliporeSigma™
712523
11 Reactions
Each for $302.00
Add to Cart
71-252-4
View Documents
MilliporeSigma™
712524
22 Reactions
Each for $537.00
Add to Cart
Description
Description
BL21 strains are the most widely used hosts for protein expression from pET recombinants and have the advantage of being deficient in both lon and ompT proteases. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
Specifications
Specifications
≤-70°C