Learn More
MilliporeSigma™ Novagen™ Rosetta-gami™ 2 Competent Cells
Rosetta-Gami 2 host strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli.
$175.00 - $346.00
Specifications
Efficiency | Guaranteed efficiency: >2 x 106 cfu/μg |
---|---|
Content And Storage | ≤-70°C |
Description
Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs.Specifications
Guaranteed efficiency: >2 x 106 cfu/μg | |
≤-70°C |