Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Novagen™ NovaBlue(DE3) Competent Cells
NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids) and recA endA mutations.
$144.00 - $299.00
Specifications
Contains F' Episome | Contains F' Episome |
---|---|
Efficiency | Guaranteed efficiency: >1 x 108 cfu/μg |
Content And Storage | ≤-70°C |
Blue/White Screening | No |
Description
NovaBlue(DE3) Competent Cells is a K-12 strain with high transformation efficiency and recA endA mutations. The DE3 lysogen of NovaBlue is potentially useful as a stringent host due to the presence of the lacIq repressor encoded by the F episome.Specifications
Contains F' Episome | |
Guaranteed efficiency: >1 x 108 cfu/μg | |
≤-70°C | |
No |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title