Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates
Receptor/ligand binding assays, sample prep,TCA precipitation
Supplier: MilliporeSigma™ MSFCN6B50
Specifications
ANSI, SBS | |
Hydrophilic PVDF | |
1.2/0.65 μm | |
Opaque | |
96 | |
0.28 cm2 | |
50/Pk. | |
1.28 cm | |
1.44 cm |
Standard | |
8 x 12 | |
Low Binding | |
Barex | |
Non-sterile | |
Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification | |
0.5 x 8.55 x 0.56 in. (1.28 x 21.71 x 1.44 cm) | |
21.71 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction