Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies
Supplier: MilliporeSigma™ MSHAN4B50
Description
- Developed for high-throughput use with automated work stations
- Individually sealed wells prevent incubation crosstalk
- Removable underdrain allows access to the filters
- Rigid sidewalls improve handling
- Ample surfaces provided for bar code labels
- Pore size: .45μm
Solid-phase RIA, DNA hybridization, ELISA and protein binding
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Standard | |
8 x 12 | |
Opaque | |
96 | |
0.28cm2 | |
50/Pk. |
Mixed Cellulose Esters | |
0.45 μm | |
Barex | |
Non-sterile | |
Solid phase RIA, DNA hybridization, ELISA, protein binding | |
8.55 cm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction