Learn More
MilliporeSigma™ Millicell-ERS2 Volt-Ohm Meter and accessories
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Supplier: MilliporeSigma™ MERS00002
Includes: Millicell-ERS2 comes with meter, battery, STX01 electrode, 1,000 Ω test electrode, battery charger. Specialized electrodes for Millicell-96 well plate are available (Cat. No. MERSSTX00).
Description
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed exclusively for use with cells cultured in Millicell™ Culture Plate Inserts.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Warranty and Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
Specifications
Millicell Ers 2V Ohm Meter | |
19 cm | |
2.3 in. | |
12.5 Hz | |
Millicell ERS-2 Voltohmmeter | |
8 to 10 hr. | |
10° to 38°C | |
1.4 kg |
7.3 in. | |
To provides qualitative measurement of cell monolayer health and quantative measures of confluency | |
6 cm | |
0 to 90% | |
1 Ea. | |
50° to 100°F | |
3 lb. | |
11 cm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.