Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Milli-Q™ Advantage A10 Ultrapure Water Purification System Accessory, Quantum Polishing Cartridges
Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements
$973.00
Specifications
For Use With (Application) | Spectrophotometry, spectroscopy, chromatography, and biochemistry |
---|---|
For Use With (Equipment) | Milli-Q™ Systems |
Catalog Number | Mfr. No. | Type | Resin Type | Purification Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Resin Type | Purification Method | Price | Quantity & Availability | |||
QTUM0TEX1
|
MilliporeSigma™
QTUM0TEX1 |
Quantum TEX Cartridge Filter | Organex & Ionex | Ion Exchange, Activated Carbon |
Each for $973.00
|
|
|||
QTUM0TIX1
|
MilliporeSigma™
QTUM0TIX1 |
Quantum TIX Cartridge Filter | Ionex | Ion Exchange |
Each for $973.00
|
|
|||
Description
Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements
Quantum™ TEX Features
- Contains Ionex resins for trace ion levels
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
- Contains Organex and Ionex resins for trace levels of organics and ions
- Feedwater nature: Ultrapure
- Technology: Activated carbon, Ion exchange
Spectrophotometry, spectroscopy, chromatography, and biochemistry
Certifications
ISO 9001 v.2000 and ISO 14001
Compliance
USP 645 suitability requirements
Specifications
Spectrophotometry, spectroscopy, chromatography, and biochemistry | |
Milli-Q™ Systems |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title