Learn More
MilliporeSigma™ MQuant™ Chlorine Test Kits
Can be used for the determination of chlorine.
Supplier: MilliporeSigma™ 1144340001
Description
The liquid reagent can be used for the determination of free and total chlorine with the MQuant Chlorine test. The chlorine concentration can be measured semiquantitatively by visual comparison of the colour of the measurement solution with the colour fields of the colour disk.Specifications
0.01 to 0.3 mg/L | |
Chlorine Test Kit |