Learn More
MilliporeSigma™ MQuant™ Chlorine Dioxide Test Kits
Used for the determination of chlorine dioxide for example in swimming pool water, drinking water and wastewater.
Supplier:  MilliporeSigma™ 1187540001
Catalog No. M1187540001
Add to Cart
Description
Description
Chlorine dioxide is used for water disinfection and as a bleaching agent in industrial processes. The test can be used for the determination of chlorine dioxide for example in swimming pool water, drinking water and wastewater. Method: colorimetric, DPD, with color card and comparator block with long test tubes 0.020 - 0.050 - 0.075 - 0.10 - 0.15 - 0.20 - 0.30 - 0.40 - 0.55mg/l ClO2 MQuant™
Specifications
Specifications
0.020 to 0.55 mg/L ClO2
Chlorine Dioxide Test Kit
Product Suggestions
Product Suggestions