Learn More
MilliporeSigma™ Upstate™ cAMP, Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 1168201ST
Description
Specifically detects cAMP Clone: in samples, and it is validated for Radioimmunoassay
Specifications
cAMP | |
Polyclonal | |
50μl serum diluted in PBS and 200μl cAMP standard in H2O. | |
2'-O-succinyl cAMP conjugated to BSA | |
RUO | |
Recognizes cAMP. Does not exhibit cross-reactivity with other nucleotides. Antibody Target Gene Symbol: CAMP Target Synonym: AntiBacteria protein Fall-39 precursor, CAP18, CATHELICIDIN, Cathelin-related antimicrobial peptide, Cnlp, CRAMP, Crp, FALL-39, hCAP-18, HSD26, MCLP Entrez Gene Name: cathelicidin antimicrobial peptide | |
≤ -70°C | |
IgG |
Radioimmune Assays (RIA) | |
Unconjugated | |
Rabbit | |
1 Set | |
Primary | |
50μL serum diluted in PBS and 200μL cAMP standard in H2O. | |
Diluted |