Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Aquaporin 3 (275-292), Rabbbit Polyclonal, ChIP Validated Antibody and Primer Set
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ 1786131ST
Description
Specifically detects Aquaporin 3 (275-292) Clone: in Rat samples, and it is validated for Immunohistochemistry
AQP3 may function as a water and urea exit mechanism in antidiuresis in collecting duct cells. It is also slightly permeable to urea and glycerol. AQP3 may play an important role in gastrointestinal tract water transport and in glycerol metabolism. AQP3 has a wide tissue distribution. In the kidney it is predominantly expressed in the inner medulla.Specifications
Aquaporin 3 (275-292) | |
Polyclonal | |
50μl antibody lyophilized from PBS, 1% BSA, 5% sucrose, pH. 7.4 and 40μg AQP3275-292 control peptide lyophilized from PBS. | |
a synthetic peptide [(KY)STEAENVKLAH(Nle)KHKEQI] corresponding to amino acids 275-292 of rat AQP3, with norleucine replacing methionine, conjugated to KLH | |
RUO | |
Recognizes the ∽40kDa and ∽29kDa forms of aquaporin-3 in rat kidney membrane. Supplied with a control peptide (immunogen). | |
Rat | |
Purified |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
1 Set | |
Primary | |
50μL antibody lyophilized from PBS, 1% BSA, 5% sucrose, pH. 7.4 and 40μg AQP3275-292 control peptide lyophilized from PBS. | |
−20°C | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction