Learn More
Thermo Scientific Dionex EGC 500 KOH Potassium Hydroxide Eluent Generator Cartridge
Supplier:  Thermo Scientific 075778
Encompass
Prevent baseline shift, increase sensitivity, improve resolution, and ensure consistent peak integration by using eluent generation. A range of Thermo Scientific™ Dionex™ EGC Cartridges are available for the production of hydroxide, carbonate, bicarbonate and methanesulfonic acid eluents. Eluent generation eliminates the need to handle acids and bases, which are traditionally required for the preparation of IC eluents, and allows chromatographers to run a full range of gradient and isocratic separations more effectively than hand-made eluents.
Catalog No. NC0959249