Learn More
Biomatik Corporation Phospho-PAK1 (Thr212) Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA51138
Sample Type: Researchers plate their cell line of choice. Species Reactivity: Human:T212, Mouse:T212, Rat:T211. Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Detection Method: Colorimetric 450 nm. Assay Time: 4.5 hours. Target Synonyms: Serine/threonine-protein kinase PAK 1; p21-activated kinase 1; PAK-1; p65-PAK; Alpha-PAK; PAK1. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.