Learn More
Biomatik Corporation Phospho-IKK-γ (Ser31) Cell-Based ELISA Kit
Supplier: Biomatik Corporation EKA51079
Sample Type: Researchers plate their cell line of choice. Species Reactivity: Human:S31. Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Detection Method: Colorimetric 450 nm. Assay Time: 4.5 hours. Target Synonyms: NEMO; NF-kappa-B essential modifier; Inhibitor of nuclear factor kappa-B kinase subunit gamma; IkB kinase subunit gamma; I-kappa-B kinase gamma; IKK-gamma; IKKG; IkB kinase-associated protein 1; IKKAP1; FIP-3. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.