Learn More
Variable Voltage Power Supply Kit
Short Protected IC Thermo.
Supplier: Elenco Electronics Inc XP15K
Description
- 1.25-15VDC 0.5A at 8V; 0.3A at12V; 0.2A at 15V
- Batteries not required
Specifications
Power Supply Kit | |
1.25/15 VDC | |
1.25 to 15vdc, 0.5a at 8v, 0.3a at 12v, 0.2a at 15v variable voltage |