Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Elements™ EM3 Polyurethane-Coated Cut-Resistant Gloves

Catalog Number p-7221491
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large

Elements™ EM3 Polyurethane-Coated Cut-Resistant knit gloves feature a waterproof membrane and polyurethane coating on the fingertips for easy use on tablets or smartphones.

  • Combines cut resistance with a waterproof membrane and nylon liner for comfort and protection
  • Membrane is constructed of a blend of nylon, metal wire and HPPE
  • Polyurethane coating on the fingertips for easy use on tablets and smartphones
  • Gloves provide ANSI Abrasion Level 2, and Cut Level A3 protection
  • Reduces the need for multiple disposable products
ANSI Abrasion Rating ANSI 2
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A3
Hand Specific
Material HPPE, Nylon, Metal
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Machining, Oily Parts Assembly, and Small Parts Handling
Glove Cuff Style Knit
Liner Material Nylon
Model EM3S
Puncture Rating ANSI/ISEA 105-16 Level 3
Series EM3
Yarn Gauge 18
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit