Learn More
National Optical Elementary Monocular Microscope
This highly portable field microscope can be used in the classroom or in the field. A great opportunity to teach microscopy on a limited budget.
Supplier: National Optical 102
Description
This highly portable field microscope can be used in the classroom or in the field. A great opportunity to teach microscopy on a limited budget.
OPTICAL SYSTEM
- 4X, 10X, and 40XR objective lenses
FOCUSING
- Intermediate range focusing
ILLUMINATION
- LED
STAGE
- Stage features locked-on, spring activated stage clips
Specifications
10X | |
LED | |
102 | |
Monocular Microscope | |
Plastic |
Monocular | |
4X, 10X, 40XR | |
Achromatic | |
Dustcover |