Learn More
Science First™ Electrostatics Demonstration Kit
SDPSCI_ED
Charge electroscopes, show electrostatic attraction/repulsion, display surface charge distribution, and more.
Supplier:  Science First™ 6153085
Catalog No. S572001
Add to Cart
Includes:
  • Two electroscopes with flask
  • Two each ball and disc terminals
  • Faraday Cage with stand and cover
  • Six different friction rods
  • Electrophorus with charge plate and handle
  • Neon lamp, aluminum ice pail, acetate cloth
  • Polyethylene film, charge transfer ball, electrically conducting ball
  • 12 pith balls, lightning rod
  • Comprehensive, 10-page instruction booklet
Description
Description
  • Teaches electrostatic conduction and induction, charge generation, charge transfer and conservation, and much more
  • Weight: 2.3kg
  • Box measures 11.5 × 11.5 × 6.5 in.
Specifications
Specifications
Teaches electrostatic conduction and induction, charge generation, charge transfer and conservation
Electrostatic Demonstration Kit
5.07 lb.
Two electroscopes with flask, two each ball and disc terminals, faraday cage with stand and cover, six different friction rods, electrophorus with charge plate and handle, Neon lamp, Aluminum ice pail, Acetate cloth, Polyethylene film, charge transfer ball, electrically conducting ball, 12 pith balls, lightning rod, instruction booklet
2.3 kg
Product Suggestions
Product Suggestions