Learn More
Research Products International Corp Disposable Electroporation Cuvettes, 4 mm, 800 ul Volume, 50 per Package
SDP
Supplier:  Research Products International Corp 248907
Encompass_Preferred
Our premium quality electroporation cuvettes provide maximum transformation efficiency. Precision molded from high strength polycarbonate to withstand high voltages. Extruded aluminum electrodes deliver a uniform pulse throughout sample for increased transformation field. Additional features include two molded side windows and snug fitting cap with frosted surface for easy marking. Fit most popular electroporation systems including BioRad, Gene Pulser and BTX. Supplied sterile in individually sealed packaging.
Catalog No. 50-212-653