Learn More
Andwin Scientific ELECTROPORATION CUVETTE 0.2 C
SDP
Supplier:  Andwin Scientific 91045050
Encompass_Preferred
The electroporation cuvette can be utilized for the electroporation of bacteria, yeast, or mammalian cells. This product is made with polished aluminum plates that minimize static and arcing for a higher quality performance. Each cuvette is wrapped individually and is gamma irradiated to eliminate the risk of cross-contamination and ensure the sanitation of the product. Color-coded caps, which signify the gap width, are included. It is sold in a case of 50.
Catalog No. 50-820-934