Learn More
Euthanex Electronic Heated Bed - Flat (10"" x 4"") (Requires EZ-HEAT Controller for operation)
Supplier: Euthanex HB1000
For use with the EZ-HEAT Surgical Bed Warming System. The warming beds ranges from 95 to 104 degrees Fahrenheit. Available in Flat (10″ x 4″ x 1″), V-shaped (10″ x 4″ x 1″) or XL size (11″ x 6″ x 1″).