Learn More
Parker Hannifan ELECTROLYTE SOLUTION
Supplier: Parker Hannifan 920071
Parker Hydrogen Generator Electrolyte, for H2PD, H2, or 75 Series. Includes (1) Bottle of Hydrogen Generator Prepared Sodium Hydroxide.