Learn More
Cole-Parmer ELECTRODE PH - HF RESISTANT DJ
Supplier:  Cole-Parmer 2711332
Encompass_Preferred
Universal sensor body, PPS construction. Models available for pH, ORP, low ionic strength solutions, and HF-resistant electrodes for solutions that should not exceed 1% hydrofluoric acid. Order pH electrode adapter at left to connect the new ERP-series electrodes to olderstyle cartridge electrode cap cable installations.User-configurable self-cleaning, flat-surfaced electrodes for almost any applicationExtended Reference Path (ERP™) technology for long life and stabilityModular plug-and-play design
Catalog No. NC2433133