Learn More
Cole-Parmer Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
Supplier: Cole-Parmer 2730119
These in-line or submersible electrodes with probe guard operate from 23 to 176 degrees Fahrenheit or -5 to 80 degrees Celsius. They have a PVC housing with a 3/4 inch NPT(M) fitting on both ends. It's epoxy body withstands pressures up to 100 psi, and measures pH from 0 to 14. ORP electrode impedance is 100 M. All electrodes measure 6-3/8 inches long by one inch outer diameter.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.