Learn More
Cole-Parmer Cole-Parmer Tuff-Tip In-Line ORP Probe, DJ/PVDF/10'/BNC
Supplier:  Cole-Parmer 0599522
Encompass_Preferred
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.
Catalog No. 50-633-485