Learn More
Cole-Parmer Oakton Ion Selective Electrode (Water hardness Ca/Mg)
Supplier: Cole-Parmer 3581248
A wide variety of ion selective electrodes are availabe to suit a wide array of applications. Select from four electrode types: membrane, solid-state, gas sensing, and glass bulb; typical response time is 20 to 30 seconds. Epoxy-body ISEs are either single- or double-junction, and the glass-body type is double junction.