Learn More
Markson Labsales ELECTRODE CLEANER SOLUTION
Supplier: Markson Labsales 62000004
Electrode Cleaner, 500 Ml. Ensure Optimum Performance From Your Ph Probe With The Electrode Cleaner. Safe For All Ph Electrodes, It Is Formulated From A Unique Combination Of Acid And Enzymes. Make It Part Of Your Ph Maintenance Program.