Learn More
Cole-Parmer Cole-Parmer Electrode storage bottle
Supplier: Cole-Parmer 0599099
Protect electrodes (up to 13 mm in diameter) against bumps, spills, and drying out. Large bottle includes one solid cap to reduce evaporation of storage solution and one O-ring cap for use with electrodes. Small bottle includes one O-ring cap and is a cost-effective option when storage solution is discarded and not regularly re-used.