Learn More
Cole-Parmer Thermo Scientific 910008-WA electrode fill solution, 3 M KCl, 5 x 60 mL bottles
Supplier: Cole-Parmer 0571925
Filling solutions for use with pH electrodes and reference electrodes. Solutions work best when combined with ionic strength and pH adjusters, which eliminate interferences from other ion species. electrode fill solution, 3 M KCl, 5 x 60 mL bottles