Learn More
Cole-Parmer Oakton Electrode Storage Solution; 500 mL
Supplier: Cole-Parmer 0065304
Keep your pH probes clean and properly stored for longevity with this storage solution that contains pepsin, removing grease and oil deposits from your electrodes. After cleaning, keep your electrodes in proper working condition with our storage solution.