Learn More
Euthanex Electronic Heated Bed, V-Shape (10"" x 4"") (Requires EZ-HEAT Controller for operation)
Supplier: Euthanex HB1000V
Electronic Heated Bed - V-Shaped (10" x 4") (Requires EZ-HEAT Controller for operation). The warming beds ranges from 95 to 104 degrees Fahrenheit. Bed temperatures can be adjusted in the administrator options.