Learn More
Cole-Parmer BB-200-115 Electric Bunsen Burner without Controller; 115 VAC
Provides clean and easy operation. Top cowl deflects heat away from hands. Air circulation from the vented housing keeps base cool to the touch. Efficient-only consumes 400 W of power.