Learn More
Vermed Inc Electrode 1-3/8" Foam Resting EKG Monitoring Echo (Case of 1000)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Vermed Inc A10009100F
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
VERMED SILVEREST Wet Gel Adult Diagnostic Electrode, 1 3/8" Foam, Metal Snap, 10/Strip, for Resting EKG and Echo Sound. LATEX-FREE, Hypoallergenic Adhesive. Ag/AgCl, high chloride content, wet gel for fast, clear traces. Self-adhering, no muscle artifact or repeat testing. Can be defibrillated.