Learn More
American Educational Products Effects of Drugs Kit
Introduces some of the physiological aspects of drugs.
Supplier: American Educational Products R101316
Description
Observe through harmless experiments the various physical effects of drugs such as aspirin, nicotine, ethyl alcohol, lactic acid, adrenaline chloride and caffeine.
- Using a culture of Daphnia, observe changes in the circulatory system and nervous activity of the organism
- Explores concepts such as stimulant/depressant, critical dosage level, reversibility of symptoms and others
- Utilizes very small amounts of drugs, so there is no danger to the students
- Class size: 30 students
Specifications
Effect of Drugs Lab Kit | |
Study the physiological aspects of drugs | |
Daphnia magna culture |
6 to 12 | |
Ringer's solution, drugs, drug dial sheet, and chambers, reproducible student worksheets and teacher's guide, msds | |
30 Students |