Learn More
Edwards Fittings for BOC High Vacuum Pumps: Short Flange
For use with BOC high vacuum pumps
$37.25 - $86.00
Description
- Made of aluminum
- Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
BOC high vacuum pump | |
Stainless Steel |