Learn More
Edvotek™ Long Wave UV Mini-Lights
Used to detect hydrolysis of fluorescent substrate.
Supplier: Edvotek™ 969
Description
- Hand-held
- Also detects fluorescent Artemia and Daphnia after their ingestion
- Can also be used for observing fluorescence in Green (gfp) and Blue (bfp) fluorescent proteins
Specifications
Handheld UV Mini Light |