Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ InstaStain™ Ethidium Bromide
Rapid and sensitive.
$95.00
Specifications
Color | Dark Red/Purple |
---|---|
Product Type | Fluorescent DNA/RNA Stain |
Stain Type | Ethidium Bromide |
Form | Sheet, Solid |
For Use With (Application) | Rapid and sensitive, disposal of InstaStain™ Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard Ethidium Bromide staining procedure |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
S05698
|
Edvotek™
2001 |
7 x 7 cm, for 40 Gels |
Each for $95.00
|
|
|||||
S05699
|
Edvotek™
2002 |
7 x 7 cm, for 100 Gels | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Contains only a few micrograms of ethidium bromide
- Gel is ready for visualization in two minutes
- Much less ethidium bromide waste for disposal compared to standard staining procedure
Specifications
Dark Red/Purple | |
Ethidium Bromide | |
Rapid and sensitive, disposal of InstaStain™ Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard Ethidium Bromide staining procedure |
Fluorescent DNA/RNA Stain | |
Sheet, Solid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title