Learn More
Edvotek™ Forensics Antigen Detection Kit
Can a deceased cat assist a murder investigation?
Supplier: Edvotek™ 192
Includes: Enough supplies for 10 student groups
Description
A young woman who lives with her cat was last seen on her daily run. After two days she was reported missing to the police by her friends. Upon entering her penthouse, police found the woman's cat in a pool of blood, shot in the head. Tiny drops of blood were found trailing from her bed sheets to the area near the cat. The detective in charge concluded that the woman and the cat were brutally murdered, the woman's body was removed from the site of the crime and the tiny stains of blood were overlooked when the area was cleaned.
Students will:
- Analyze samples that were obtained from the “blood” around the dead cat
- Determine if the blood stains on the bed sheets are human or feline
- Determine the validity of the hypothesis set forth by the detective
No actual blood or blood products used in this experiment.
Specifications
Determine human vs animal source of blood, evaluate hypothesis of fictional detective | |
Forensics Antigen Detection Kit | |
Refrigerated |
Instructions, simulated control and crime scene blood samples, antigen/antibody detection reagents, microtiter plates, ultraspec agarose™, practice loading solution, petri plates, well cutters | |
Distilled or deionized water, beakers, 37°C Incubation oven, disposable lab gloves, safety goggles, automatic micropipets (100μL) and tips recommended, plastic container, plastic wrap, pipets - 5 or 10mL, marking pen | |
10 Lab Groups |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.