Learn More
Edvotek™ Electrophoretic Properties of Native Proteins
Proteins are complex biomolecules with varying charge, size and shape that can be analyzed by agarose gel electrophoresis.
Supplier: Edvotek™ 111
Includes:
- Instructions
- Protein samples
- Gel-loading solution
- Agarose
- Electrophoresis buffer
- Stain
Requires :
- Horizontal electrophoresis apparatus
- Power supply
- Visualization (white light)
- Automatic micropipet with tips
- Microwave or hotplate
- Water bath
- Metric rulers
- Lab glassware
- Methanol
- Glacial acetic acid
- Distilled or deionized water
Description
- Students use gel analysis of native proteins to evaluate natural charge and shape characteristics of proteins
- After electrophoresis, the protein samples are stained for visualization
Experiment times:
- Gel prep, 30 minutes
- Electrophoresis, 45 minutes
- Staining, one hour
- Destaining, overnight
Order Info
Contains materials for six gels.
Specifications
Electrophoresis Kit | |
Instructions, protein samples, gel loading solution, agarose, electrophoresis buffer, protein insta stain | |
45 minutes; Staining time: 60 minutes |
Evaluate natural charge and shape characteristics of proteins | |
6 Gels | |
Refrigerated; Store at: 4°C; 39.2°F |