Learn More
Malvern Panalytical Edta/Caci Test Kit Vp-Itc
Supplier: Malvern Panalytical KIT240010
EDTA Test Kit VP-ITC. This kit allows users to test instrument performance using EDTA. Expect material for least 3 single use runs from this kit. This kit will ship with at least 90 days of use before material expiry.