Learn More
Justrite™ EcoPolyBlend™ Accumulation Center
Create custom systems to deliver a complete solution. The Justrite™ EcoPolyBlend™ Accumulation Center combines units to create customized spill-control systems, allowing for the perfect solution to specific industry needs.
$775.87 - $1250.17
Catalog Number | Mfr. No. | Holds | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Color | Price | Quantity & Availability | ||||
19-822-314
|
Justrite™
28658 |
6 Drums | Yellow |
Each for $836.59
|
|
||||
19-822-313
|
Justrite™
28659 |
6 Drums | Black |
Each for $775.87
|
|
||||
19-822-316
|
Justrite™
28660 |
8 Drums | Yellow |
Each for $1,250.17
|
|
||||
19-822-315
|
Justrite™
28661 |
8 Drums | Black |
Each for $1,220.25
|
|
||||
Description
Low-Profile EcoPolyBlend™ Accumulation Centers only measure 5.5 in. (140-mm) high for easy handling of drums. Made of recycled polyethylene, these accumulation centers are nearly impervious to chemicals, including acids and corrosives. Seamless construction eliminates leaks, making these ideal for drum filling and dispensing operations. Self-locating grates won’t ride-up, and are removable for easy cleaning. Combine 2-, 4-, 6-, and 8-drum modular units to create your own customized system. Use the optional joining clips to firmly lock units together. Sumps remain separate and clips can be removed for easy repositioning.Spot an opportunity for improvement?