Learn More
Mystaire™ Economy Workstation Main Filter
For use with the Mystaire™ Economy Workstation (FE-2620). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Supplier: Mystaire™ FE1021J
Description
- Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
- Made in the U.S.A.
- Each filter application is reviewed to ensure a proper filtration package is selected.
- Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
FE-2620 | |
Filter | |
Gas Phase Carbon | |
Mineral Acids |