Learn More
World Precision Instrument Economy Tweezers #7, 11cm, Curved, 0.4x0.5mm Tips
Supplier: World Precision Instrument 501981
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.