Learn More
Cole-Parmer Cole-Parmer Economical Square HDPE Carboy w/ Handle, 10 L (2.5 Gal), 1/ea
Supplier: Cole-Parmer 0603180
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Durable, leakproof square carboys save space. Large carrying handles and an indented bottom make handling and pouring easy. Molded HDPE threaded closure included. Can be drilled to accept a 3/4'' faucet, sold separately. Temperature range: 68 to 230F (20 to 110C).