Learn More
Zymo Research Corporation E. coli Non-Methylated Genomic DNA (5 μg/ 20 μl)
Supplier: Zymo Research Corporation D5016
E. coli Non-methylated Genomic DNA for use as a negative control for epigenetic experiments requiring DNA containing zero background levels of cytosine and adenosine methylation. Substrate for DNA methylation-sensitive restriction enzyme digestions.