Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZING™ Enterprises Eco GHS Projecting Sign, SDS
ZING Enterprises Eco Safety sign available in V-Style, L-Style, Standard TriView, and Slim Triview signage.
Supplier: ZING™ Enterprises 6051
Description
Features:
- Identifies designated SDS information areas
- Supports compliance with the new GHS/Hazard Communication requirements.
- Significantly increases visibility of information
- Made from high quality 70% post-consumer recycled plastic
- UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
- Recycled material supports sustainability initiatives and LEED credits
- Includes 2 mounting holes and mounting adhesive tabs and measures.
Certifications
GHS OSHA Hazard Communication 2012
Specifications
Plastic | |
Corner Holes | |
7.5 in. | |
English | |
SDS |
1 Ea. | |
Communication | |
20 in. | |
7.5 x 20 in. | |
Red on White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction