Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises Eco GHS Projecting Sign, SDS

ZING Enterprises Eco Safety sign available in V-Style, L-Style, Standard TriView, and Slim Triview signage.

Supplier:  ZING™ Enterprises 6050

 View more versions of this product

Catalog No. 18-001-875


Add to Cart

Description

Description

Features:

  • Identifies designated SDS information areas
  • Supports compliance with the new GHS/Hazard Communication requirements.
  • Significantly increases visibility of information
  • Made from high quality 70% post-consumer recycled plastic
  • UL Validated for recycled content which substantiates material compliance with various green codes, standards, and procurement policies including the LEED Green Building Rating System, IgCC, GSA Advantage, ASHRAE 189.1 and others.
  • Recycled material supports sustainability initiatives and LEED credits
  • Includes 2 mounting holes and mounting adhesive tabs and measures.

Certifications

GHS OSHA Hazard Communication 2012

Specifications

Specifications

Plastic
Corner Holes
7 in.
English
SDS
1 Ea.
Communication
2.5 in.
7 x 2.5 in.
Red on White
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit