Learn More
AVANTOR MASTERFLEX (VWR LLC) Flowline DL14-01 EchoPod Level Switch with FOB USB Interface
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0750603
Encompass_Preferred
This level switch is an integrated control automation solution to provide noncontact level measurement with switch and control capabilities. Its compact design allows for installation in applications with limited mounting options and space. It measures dirty, sticky and corrosive media without affecting product performance, and an advanced digital signal processing provides surface imaging in applications with obstacles in the beam width. A single unit can provide switch, control and measurement capabilities simultaneously or individually.
Catalog No. 50-630-862
Explore available promotions