Learn More
Hanna Pharmaceutical Supply Co., Inc. Electrode Cream Signacreme 5oz. Squeeze Bottle
Supplier: Hanna Pharmaceutical Supply Co., Inc. 1705
Highly conductive cream electrolyte for use in electromedical procedures where a cosmetic quality cream is preferred Cosmetic quality, pleasing to patients Non-irritating, hypoallergenic, bacteriostatic Conveniently packaged in refillable see-thru dispenser Flip-Top Cap Not recommended for defibrillation
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.